Order Entry
Netherlands
ContactUsLinkComponent
VWR®, Standard solution (1000 ppm NO₃) for the preparation of nitrate standard solution (100 ppm NO₃) Reag. Ph. Eur.; 5002100
VWR®, Standard solution (1000 ppm NO₃) for the preparation of nitrate standard solution (100 ppm NO₃) Reag. Ph. Eur.; 5002100
Catalog # 88088.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Standard solution (1000 ppm NO₃) for the preparation of nitrate standard solution (100 ppm NO₃) Reag. Ph. Eur.; 5002100
Catalog # 88088.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Nitrate standard solution
  • Contents:
    KNO₃ in H₂O

Specifications

About this item

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.