
4'-Carboxybenzo-18-crown-6 (4'-carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:60835-75-8
:
Formule:C₁₇H₂₄O₈
MDL-nummer:MFCD00143277
:356.3677 g/mol
:GHS07
Specificatie testresultaten
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:123.0 to 127.0 deg-C
Specificatie testresultaten
Gerelateerde informatie
Frequently Bought Together