Methyl-3-methyl-2-nitrobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5471-82-9
:
Formula:C₉H₉NO₄
MDL Number:MFCD01003947
:195.17 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:72.0 to 76.0 deg-C
Specification Test Results
Frequently Bought Together