

N,N'-Dimethylhydrazine dihydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAD0741-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
Merck Index:14,03248
EINECS:206-183-5
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:306-37-6
Formula:C₂H₁₀Cl₂N₂
MDL Number:MFCD00012522