Order Entry
Netherlands
ContactUsLinkComponent
(±)-2,3-Epoxypropyl methacrylate ≥95.0% (by GC) stabilised
Corrosive
Toxic
Health
(±)-2,3-Epoxypropyl methacrylate ≥95.0% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
Corrosive
Toxic
Health
(±)-2,3-Epoxypropyl methacrylate ≥95.0% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together