Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dimethylcyclohexanol (mixture of isomers) ≥99.0% (by GC)
Flammable
2,6-Dimethylcyclohexanol (mixture of isomers) ≥99.0% (by GC)
Catalog # TCIAD1578-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
2,6-Dimethylcyclohexanol (mixture of isomers) ≥99.0% (by GC)
Catalog # TCIAD1578-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 % (total of isomers)

Specification Test Results

About this item

 :
Boiling Pt:173°C
Flash Pt:55°C
EINECS:226-264-9
UN:1987
ADR:3,III
 :GHS02
CAS Number:5337-72-4
Formula:C₈H₁₆O
MDL Number:MFCD00001502