

(R)-(-)-Glycidyl butyrate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:60456-26-0
:
Formula:C₇H₁₂O₃
MDL Number:MFCD00075120
:144.17 g/mol
Boiling Pt:90°C
Flash Pt:84°C
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:-29.0 to -33.0 deg (neat)
Specification Test Results
Frequently Bought Together