

2-Butyl-5-chloroimidazole-4-carbaldehyde ≥98.0% (by GC)
Catalog # TCIAB2914-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:96.0 to 100.0 deg-C
Specification Test Results
About this item
:
:186.6387 g/mol
EINECS:410-260-0
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:83857-96-9
Formula:C₈H₁₁ClN₂O
MDL Number:MFCD01934396