

(R)-(-)-Glycidyl methyl ether ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:64491-70-9
:
Formula:C₄H₈O₂
MDL Number:MFCD00273372
UN:1992
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Specific rotation [a]20/D:-12.0 to -15.0 deg (Neat)
Specification Test Results
Frequently Bought Together