
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:16178-48-6
:
Formula:C₁₀H₁₃N₅Na₂O₁₀P₂·2H₂O
MDL Number:MFCD00066635
:471.167 g/mol (anhy)
EINECS:240-314-7
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 93.0 area%
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-29.0 to -32.0 deg (C=0.5, 0.5mol/L Na2HPO4 sol.)
- Water:max. 10.0 %
Specification Test Results
Frequently Bought Together