

Cyclopropanemethylamine ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2516-47-4
:
Formula:C₄H₉N
MDL Number:MFCD00037147
:71.12 g/mol
Boiling Pt:86°C
Density:0,82 g/cm³ (20 °C)
Flash Pt:2°C
Storage Temperature:Ambient
EINECS:219-737-6
UN:2733
ADR:3,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together