Order Entry
Netherlands
ContactUsLinkComponent
Sodium benzoate, Supelco®
Sodium benzoate, Supelco®
Catalog # PHR1231-1G
Supplier:  Merck
CAS Number:  
undefined
Sodium benzoate, Supelco®
Catalog # PHR1231-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Benzoic acid sodium salt
 :
 :144.11 g/mol
Melting Pt:410…430 °C
Density:1.44 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:208-534-8
REACH:01-2119460683-35
CAS Number:532-32-1
Formula:C₆H₅COONa
MDL Number:MFCD00012463
Merck Index:13,08654