Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
2-Fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
 
2-Fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow to Light orange powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

Frequently Bought Together