
3-Chloro-2-fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:351003-48-0
:
Formula:C₆H₃Cl₂FO₂S
MDL Number:MFCD03094234
:229.06 g/mol
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together