Order Entry
Netherlands
ContactUsLinkComponent
cis-1-(1-Ethoxyethoxy)-3-hexene ≥97.0% (by GC)
cis-1-(1-Ethoxyethoxy)-3-hexene ≥97.0% (by GC)
Catalog # TCIAA0928-25ML
Supplier:  TCI
CAS Number:  
undefined
cis-1-(1-Ethoxyethoxy)-3-hexene ≥97.0% (by GC)
Catalog # TCIAA0928-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
Flash Pt:70°C
 :172.27 g/mol
EINECS:248-817-3
CAS Number:28069-74-1
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00036618