Order Entry
Netherlands
ContactUsLinkComponent
(2-Chlorobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC)
Irritant
(2-Chlorobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC)
Catalog # TCIAC1759-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(2-Chlorobenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC)
Catalog # TCIAC1759-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
Melting Pt:244°C
 :423.3140 g/mol
 :GHS07
CAS Number:18583-55-6
Formula:C₂₅H₂₁Cl₂P
MDL Number:MFCD00054629