Order Entry
Netherlands
ContactUsLinkComponent
cis-3-Hexenyl acetate ≥97.0% (by GC)
Flammable
cis-3-Hexenyl acetate ≥97.0% (by GC)
Catalog # TCIAA0888-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
cis-3-Hexenyl acetate ≥97.0% (by GC)
Catalog # TCIAA0888-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :142.20 g/mol
Boiling Pt:169°C
Flash Pt:57°C
EINECS:222-960-1
UN:3272
ADR:3,III
 :GHS02
CAS Number:3681-71-8
Formula:C₈H₁₄O₂
MDL Number:MFCD00036563