(R)-4-Benzyloxy-1,3-butanediol ≥96.0% (by GC)
Catalog # TCIAB2901-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 96.0 %
- Optical purity (LC):min. 98.0 ee%
- Specific rotation [a]20/D:+7.0 to +13.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
Boiling Pt:138°C
Storage Temperature:Refrigerator
CAS Number:81096-93-7
Formula:C₁₁H₁₆O₃
MDL Number:MFCD11846117