Bestelling invoeren
Netherlands
ContactUsLinkComponent
2'-Methoxyacetanilide ≥98.0% (by HPLC, total nitrogen)
Irritant
2'-Methoxyacetanilide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA0018-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2'-Methoxyacetanilide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA0018-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    85.0 to 89.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :165.1891 g/mol
EINECS:202-233-5
 :GHS07
CAS Number:93-26-5
Formula:C₉H₁₁NO₂
MDL Number:MFCD00026117