


Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:139-08-2
:
Formula:C₂₃H₄₂ClN·H₂O
MDL Number:MFCD00011771
:368.05 g/mol (anhy)
Merck Index:14,01059
EINECS:205-352-0
UN:2923
ADR:8,III
:GHS05
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 95.0 % (as C14)
- Purity (TPBNa Method):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 12.0 %
Specification Test Results
Frequently Bought Together