Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Tributyldodecylphosphonium bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAD2912-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Tributyldodecylphosphonium bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAD2912-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :451.5475 g/mol
 :GHS07
CAS Number:15294-63-0
Formula:C₂₄H₅₂BrP
MDL Number:MFCD04038166