
trans-2-Butene-1,4-bis(triphenylphosphonium chloride) ≥95.0% (by titrimetric analysis)
Catalog # TCIAB2286-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 95.0 %
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
About this item
:
:649.60 g/mol
:GHS07
CAS Number:106423-29-4
Formula:C₄₀H₃₆Cl₂P₂
MDL Number:MFCD00031713