



4-Fluorphenylmagnesium bromide ∼19% (w/w) ∼1.0 mol/L in THF
TCIAF0673-250G
: TCI
:
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
- Concentration (sec Butanol method):18.5 to 20.5 w/w%
:
:-20°C
:2924
:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:352-13-6
:C₆H₄BrFMg
:MFCD00009667
:–20°C