Order Entry
Netherlands
ContactUsLinkComponent
2-Methoxy-6-(methylamino)pyridine ≥98.0% (by GC, titration analysis)
Irritant
2-Methoxy-6-(methylamino)pyridine ≥98.0% (by GC, titration analysis)
Catalog # TCIAM1185-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Methoxy-6-(methylamino)pyridine ≥98.0% (by GC, titration analysis)
Catalog # TCIAM1185-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :138.17 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:88569-83-9
Formula:C₇H₁₀N₂O
MDL Number:MFCD00191557