
1,4-Butandiol monovinyl ether ≥97.0% (by GC) stabilised
: TCI
:17832-28-9
:
:C₆H₁₂O₂
:MFCD00080697
:116.16 g/mol
:86°C
:80°C
:Stabilised with potassium hydroxide
:241-793-5
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together