L(-)-Arabitol
Catalog # EHERC10298000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:152.15 g/mol
EINECS:231-582-6
CAS Number:7643-75-6
Formula:C₅H₁₂O₅
MDL Number:MFCD00064290