Order Entry
Netherlands
ContactUsLinkComponent
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide ≥97.0% (by titrimetric analysis)
Corrosive
Irritant
Health
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
Health
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together