
2'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:92-15-9
:
Formula:C₁₁H₁₃NO₃
MDL Number:MFCD00008781
:207.2300 g/mol
EINECS:202-131-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:85.0 to 87.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together