Order Entry
Netherlands
ContactUsLinkComponent
Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥97.0% (by GC) stabilised
Irritant
Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥97.0% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
Irritant
Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥97.0% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
  • Acid Value:
    max. 0.5

Specification Test Results

Related Information

Frequently Bought Together