Order Entry
Netherlands
ContactUsLinkComponent
4-Chloromethyl-m-xylene ≥80.0% (by GC) (contains 2,6-isomer)
Corrosive
4-Chloromethyl-m-xylene ≥80.0% (by GC) (contains 2,6-isomer)
Catalog # TCIAD0288-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
4-Chloromethyl-m-xylene ≥80.0% (by GC) (contains 2,6-isomer)
Catalog # TCIAD0288-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 80.0 %

Specification Test Results

About this item

 :
Boiling Pt:105°C
Density:1,058 g/cm³ (19 °C)
EINECS:212-531-7
UN:3265
ADR:8,II
CAS Number:824-55-5