
4-Chloro-3-fluoroaniline ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:367-22-6
:
Formula:C₆H₅ClFN
MDL Number:MFCD01090987
:145.56 g/mol
Boiling Pt:226°C
EINECS:206-683-3
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:58.0 to 62.0 deg-C
Specification Test Results
Frequently Bought Together