Order Entry
Netherlands
ContactUsLinkComponent
1,1-Dibromoethane ≥98.0% (by GC) stabilised
Toxic
1,1-Dibromoethane ≥98.0% (by GC) stabilised
Catalog # TCIAD0184-5G
Supplier:  TCI
CAS Number:  
undefined
Toxic
1,1-Dibromoethane ≥98.0% (by GC) stabilised
Catalog # TCIAD0184-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :187.86 g/mol
Boiling Pt:108°C
Melting Pt:–0.5 °C
Density:2,10294 g/cm³ (15 °C)
EINECS:209-184-9
UN:2810
ADR:6.1,III
 :Stabilised with copper chip
 :GHS06
CAS Number:557-91-5
Formula:Br₂HCCH₃
MDL Number:MFCD00041719