
Sinapic acid ≥99.0% (by GC, titration analysis) matrix for MALDI-TOF/MS
TCIAD2932-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
:
:224.21 g/mol
:378 °C (1013 hPa)
:202°C
:208-487-3
:GHS07
:530-59-6
:C₁₁H₁₂O₅
:MFCD00004401