Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, Disulfoton 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Disulfoton 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL12980000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Disulfoton 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL12980000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    298-04-4
  • Description:
    Disulfoton 10 µg/ml in cyclohexane
  • Formula:
    C₈H₁₉O₂PS₃
  • Description:
    Disulfoton

Specifications

About this item