Calcium perchlorate hydrate (12,2 - 13,5% Ca), Reagent Grade
Catalog # 11655.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Verpakking:Glass bottle for solids
- VPE:250 g
- Seq:3545
Specifications
About this item
:
:238.98 g/mol (anhy)
Storage Temperature:Kamertemperatuur
EINECS:236-768-0
UN:1455
ADR:5.1,II
:GHS03
:GHS07
CAS Number:13477-36-6
Formula:Ca(ClO₄)₂·xH₂O
MDL Number:MFCD00015971