1,3-Di-tert-butyltoluene ≥98.0% (by GC)
: TCI
:15181-11-0
:
:C₁₅H₂₄
:MFCD00026300
:244°C
:204.36 g/mol
:239-230-3
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
Frequently Bought Together