Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
 
2,4'-Dimethoxybenzophenone ≥98.0% (by GC)
  TCIAD2890-5G
 :  TCI
 :  
 
2,4'-Dimethoxybenzophenone ≥98.0% (by GC)
  TCIAD2890-5G
 :  TCI
 :  

 

  • Verpakking:
    Glass bottle
  • VPE:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White or Colorless powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    96.0 to 100.0 deg-C
  • Solubility in Toluene:
    almost transparency

 

 

 :
 :GHS07
 :5449-69-4
 :C₁₅H₁₄O₃
 :MFCD01311508