L-(-)-Fucose ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:2438-80-4
:MFCD00135607
Formula:C₆H₁₂O₅
:164.16 g/mol
Merck Index:14,04279
EINECS:219-452-7
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC) (CAD):min. 97.0 area%
- Melting point:135.0 to 139.0 deg-C
- Specific rotation [a]20/D:-72.0 to -77.0 deg (C=1, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together