L-(-)-Fucose 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2438-80-4
:MFCD00135607
Formula:C₆H₁₂O₅
MDL Number:MFCD00069812
:164.16 g/mol
Melting Pt:128…141 °C
Storage Temperature:Ambient
EINECS:219-452-7
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- HPLC:≥96.0 % (ELSD)
- Specific optical rotation:-72° to -78° (20°C, 589 nm) (c=4, H2O, NH3)
Specification Test Results
Frequently Bought Together