1,3-Di(o-tolyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD0802-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:149.0 to 158.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:256.37 g/mol
EINECS:205-309-6
CAS Number:137-97-3
Formula:C₁₅H₁₆N₂S
MDL Number:MFCD00025922