2-(2,4,5-Trichlorophenoxy)propionic acid
Catalog # EHERC13490000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:269.51 g/mol
EINECS:202-271-2
CAS Number:93-72-1
Formula:C₉H₇Cl₃O₃
MDL Number:MFCD00002646