Order Entry
Netherlands
ContactUsLinkComponent
2,2,2-Trifluoroethyl trifluoromethanesulphonate ≥97.0% (by GC)
Flammable
Corrosive
Irritant
Health
2,2,2-Trifluoroethyl trifluoromethanesulphonate ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
2,2,2-Trifluoroethyl trifluoromethanesulphonate ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together