



Sodium bis(trimethylsilyl)amide 38% ∼1.9 mol/L in THF (contains 2-Methyl-2-butene stabilised
: TCI
:1070-89-9
:
:C₆H₁₈NNaSi₂
:MFCD00009835
:183.38 g/mol
:3,II
:Stabilised with 2-methyl-2-butene (iso-amylene)
:213-983-8
:2924
:GHS02
:GHS05
:GHS07
:GHS08
:Ambient
- Appearance:Light yellow to Amber to Dark green clear liquid
- Concentration (Nonaqueous Titration):38.0 to 42.5 w/w%
Frequently Bought Together