
3,4-Dihydroxybenzophenone ≥98.0% (by GC, titration analysis)
: TCI
:10425-11-3
:
:C₁₃H₁₀O₃
:MFCD00477203
:214.2200 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:146.0 to 150.0 deg-C
Frequently Bought Together