
Di(2-pyridyl) ketone ≥98.0% (by titrimetric analysis)
TCIAD1708-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Orange to Green powder to crystaline
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:52.0 to 56.0 deg-C
:
:184.20 g/mol
:243-067-3
:GHS07
:19437-26-4
:C₁₁H₈N₂O
:MFCD00006288