Order Entry
Netherlands
ContactUsLinkComponent
Disulfoton-[diethyl-D10] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Disulfoton-[diethyl-D10] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12980100CY
Supplier:  EHRENSTORFER
CAS Number:  
Disulfoton-[diethyl-D10] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12980100CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,1 mL
  • CAS No.:
    298-04-4
  • Description:
    Disulfoton-[diethyl-D10] 100 µg/ml in cyclohexane
  • Formula:
    C₈H₁₉O₂PS₃
  • Description:
    Disulfoton-[diethyl-D10]

Specifications

About this item