3-Bromobenzaldehyde ethylene acetal ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17789-14-9
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00003209
:229.07 g/mol
EINECS:241-766-8
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together