
2-Bromobenzaldehyde ethylene acetal ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34824-58-3
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00155124
:229.07 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together