
Benzophenone-2,4,5-tricarboxylic acid ≥98.0% (by titrimetric analysis)
TCIAB2421-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:very faint turbidity
:
:314.246 g/mol
:GHS07
:135989-69-4
:C₁₆H₁₀O₇
:MFCD08276278