
5-Chloro-2-(methylamino)benzophenone ≥98.0% (by GC)
TCIAC2281-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:94.0 to 99.0 deg-C
:
:245.71 g/mol
:213-822-1
:GHS07
:1022-13-5
:C₁₄H₁₂ClNO
:MFCD00008284